1-Ethynyl-2,4-difluorobenzene ≥96.0%

Leverantör: TCI EUROPE
Varning

E1169-1G E1169-5G
TCIAE1169-1GEA 1900 SEK
TCIAE1169-1G TCIAE1169-5G
1-Ethynyl-2,4-difluorobenzene ≥96.0%
1-Ethynyl-2,4-difluorobenzene
Formel: C₈H₄F₂
MW: 138,12 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD02093726
CAS nummer: 302912-34-1
UN: 1325
ADR: 4.1,III

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR