4-Ethyl-4-methylmorpholinium bromide ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Ethyl-4-methylmorpholin-4-ium bromide
E1166-5G
E1166-25G
TCIAE1166-5GEA
1050
SEK
TCIAE1166-5G
TCIAE1166-25G
4-Ethyl-4-methylmorpholinium bromide ≥97.0%
4-Ethyl-4-methylmorpholinium bromide
Formel:
C₇H₁₆BrNO MW: 210,12 g/mol |
MDL Number:
MFCD03095385 CAS nummer: 65756-41-4 EINECS: 418-250-8 |
Specifikation testresultat
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Melting point | 177.0 to 187.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...