4-Ethynyl-1,1'-biphenyl ≥98.0%

Leverantör: TCI

E1141-1G E1141-5G
TCIAE1141-1GEA 565 SEK
TCIAE1141-1G TCIAE1141-5G
4-Ethynyl-1,1'-biphenyl ≥98.0%
4-Ethynyl-1,1'-biphenyl
Formel: C₁₄H₁₀
Storage Temperature: Kylskåp
MDL Number: MFCD00102191
CAS nummer: 29079-00-3

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 86.0 to 90.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR