Ethoxy(pentafluoro)cyclotriphosphazene ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 2-Ethoxy-2,4,4,6,6-pentafluoro-1,3,5,2,4,6-triazatriphosphorine

E1140-1G E1140-5G
TCIAE1140-1GEA 830 SEK
TCIAE1140-1G TCIAE1140-5G
Ethoxy(pentafluoro)cyclotriphosphazene ≥98.0%
Ethoxy(pentafluoro)cyclotriphosphazene
Formel: C₂H₅F₅N₃OP₃
MW: 275,00 g/mol
Kokpunkt: 42°C
Storage Temperature: Kylskåp
MDL Number: MFCD28386107
CAS nummer: 33027-66-6
UN: 1760
ADR: 8,III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR