3-Ethoxybenzaldehyde ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
E1133-5G
E1133-25G
TCIAE1133-5GEA
525
SEK
TCIAE1133-5G
TCIAE1133-25G
3-Ethoxybenzaldehyde ≥98.0%
3-Ethoxybenzaldehyde
Formel:
C₉H₁₀O₂ MW: 150,18 g/mol Kokpunkt: 245°C Flampunkt: 110°C |
MDL Number:
MFCD00016606 CAS nummer: 22924-15-8 EINECS: 245-333-4 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...