Ethyl-2-formyl-3-oxopropionate ≥97.0%

Leverantör: TCI
Varning

E1128-1G E1128-5G
TCIAE1128-1GEA 1025 SEK
TCIAE1128-1G TCIAE1128-5G
Ethyl-2-formyl-3-oxopropionate ≥97.0%
Ethyl-2-formyl-3-oxopropionate
Formel: C₆H₈O₄
Kokpunkt: 32°C
Storage Temperature: Kylskåp
MDL Number: MFCD11112084
CAS nummer: 80370-42-9

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR