4-Ethoxy-3-fluorophenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI
Varning

E1124-1G E1124-5G
TCIAE1124-1GEA 215 SEK
TCIAE1124-1G TCIAE1124-5G
4-Ethoxy-3-fluorophenylboronic acid (contains varying amounts of Anhydride)
4-Ethoxy-3-fluorophenylboronic acid
Formel: C₈H₁₀BFO₃
MW: 183.97 g/mol
MDL Number: MFCD04115667
CAS nummer: 279263-10-4

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 111.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR