Ethyl-5-bromothiophene-2-carboxylate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Ethyl 5-bromothiophene-2-carboxylate

E1111-1G E1111-5G
TCIAE1111-1GEA 350 SEK
TCIAE1111-1G TCIAE1111-5G
Ethyl-5-bromothiophene-2-carboxylate ≥98.0%
Ethyl-5-bromothiophene-2-carboxylate
Formel: C₇H₇BrO₂S
MW: 235,10 g/mol
Kokpunkt: 175°C
MDL Number: MFCD02683089
CAS nummer: 5751-83-7

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR