Ethyl-4,6-dihydroxynicotinate ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Ethyl 4,6-dihydroxynicotinate

E1110-1G E1110-5G
TCIAE1110-1GEA 455 SEK
TCIAE1110-1G TCIAE1110-5G
Ethyl-4,6-dihydroxynicotinate ≥97.0%
Ethyl-4,6-dihydroxynicotinate
Formel: C₈H₉NO₄
MW: 183,17 g/mol
MDL Number: MFCD00834976
CAS nummer: 6975-44-6

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR