Ethyl-3-(benzylamino)-3-oxopropionate ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Ethyl N-Benzylmalonamate, Ethyl (N-Benzylcarbamoyl)acetate, N-Benzylmalonamic Acid Ethyl Ester, (N-Benzylcarbamoyl)acetic Acid Ethyl Ester, 3-(Benzylamino)-3-oxopropionic Acid Ethyl Ester, Ethyl 3-(benzylamino)-3-oxopropanoate
E1098-1G
E1098-5G
TCIAE1098-1GEA
1475
SEK
TCIAE1098-1G
TCIAE1098-5G
Ethyl-3-(benzylamino)-3-oxopropionate ≥98.0%
Ethyl-3-(benzylamino)-3-oxopropionate
Formel:
C₁₂H₁₅NO₃ MW: 221,26 g/mol |
MDL Number:
MFCD28128094 CAS nummer: 29689-63-2 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 49.0 to 53.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...