(-)-Epigallocatechin ≥98.0%

Leverantör: TCI EUROPE

Synonyms: (2R,3R)-2-(3,4,5-Trihydroxyphenyl)chromane-3,5,7-triol, 3,5,7,3',4',5'-Hexahydroxylflavan

E1084-10MG E1084-50MG
TCIAE1084-10MGEA 1175 SEK
TCIAE1084-10MG TCIAE1084-50MG
(-)-Epigallocatechin ≥98.0%
(-)-Epigallocatechin
Formel: C₁₅H₁₄O₇
MW: 306,27 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00075939
CAS nummer: 970-74-1

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -48.0 to -51.0 deg (C=0.5, Acetone)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR