2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Ethynylboronic acid Pinacol ester

E1074-1G E1074-200MG
TCIAE1074-1GEA 2375 SEK
TCIAE1074-1G TCIAE1074-200MG
2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥95.0%
2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel: C₈H₁₃BO₂
MW: 152,00 g/mol
Kokpunkt: 140°C
Storage Temperature: Kylskåp
MDL Number: MFCD13182122
CAS nummer: 347389-74-6
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b8424551b6ea23aac6180100e57fcb709444cbc&vid=3b8424551b6ea23aac6180100e57fcb709444cbc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR