Ethyl-4,6-dichloronicotinate ≥98.0%
Leverantör: TCI
|
Varning
|
Synonyms:
Ethyl 4,6-dichloronicotinate
E1068-1G
E1068-5G
TCIAE1068-1GEA
470
SEK
TCIAE1068-1G
TCIAE1068-5G
Ethyl-4,6-dichloronicotinate ≥98.0%
Ethyl-4,6-dichloronicotinate
Formel:
C₈H₇Cl₂NO₂ MW: 220.05 g/mol Kokpunkt: 85°C |
MDL Number:
MFCD00173839 CAS nummer: 40296-46-6 |
Specifikation testresultat
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 31.0 to 35.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...