9-(4-Ethynylphenyl)carbazole ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
|
Varning
|
E1055-1G
E1055-5G
TCIAE1055-1GEA
1300
SEK
TCIAE1055-1G
TCIAE1055-5G
9-(4-Ethynylphenyl)carbazole ≥98.0% (by HPLC, total nitrogen)
9-(4-Ethynylphenyl)carbazole
Formel:
C₂₀H₁₃N MW: 267,33 g/mol |
MDL Number:
MFCD12024288 CAS nummer: 262861-81-4 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 104.0 to 108.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...