1-Ethyl-1-methylpyrrolidinium bromide ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1-Ethyl-1-methylpyrrolidin-1-ium bromide

E1050-5G E1050-25G
TCIAE1050-5GEA 215 SEK
TCIAE1050-5G TCIAE1050-25G
1-Ethyl-1-methylpyrrolidinium bromide ≥97.0%
1-Ethyl-1-methylpyrrolidinium bromide
Formel: C₇H₁₆BrN
MW: 194,12 g/mol
MDL Number: MFCD03095384
CAS nummer: 69227-51-6
EINECS: 418-200-5

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR