Eslicarbazepine acetate ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
BIA 2-093, (S)-(-)-10-Acetoxy-10,11-dihydro-5H-dibenz[b,f]azepine-5-carboxamide
E1046-200MG
E1046-1G
TCIAE1046-200MGEA
1500
SEK
TCIAE1046-200MG
TCIAE1046-1G
Eslicarbazepine acetate ≥98.0% (by HPLC, total nitrogen)
Eslicarbazepine acetate
Formel:
C₁₇H₁₆N₂O₃ MW: 296,33 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD06798333 CAS nummer: 236395-14-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 185.0 to 189.0 deg-C |
Specific rotation [a]20/D | -20.0 to -24.0 deg (C=1, Pyridine) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...