Ethyl 4-Bromo-3-methylbenzoate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Ethyl 4-bromo-3-methylbenzoate, 4-Bromo-m-toluic acid Ethyl ester, 4-Bromo-3-methylbenzoic acid Ethyl ester, Ethyl 4-Bromo-m-toluate

E1036-5G E1036-25G
TCIAE1036-5GEA 605 SEK
TCIAE1036-5G TCIAE1036-25G
Ethyl 4-Bromo-3-methylbenzoate ≥98.0%
Ethyl 4-Bromo-3-methylbenzoate
Formel: C₁₀H₁₁BrO₂
MW: 243,10 g/mol
Kokpunkt: 275°C
MDL Number: MFCD00673007
CAS nummer: 160313-69-9

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR