2-Ethyl-1-hexanethiol ≥98.0%

Leverantör: TCI
Varning

Synonyms: 2-Ethylhexyl Mercaptan

E1031-25G E1031-100G
TCIAE1031-25GEA 765 SEK
TCIAE1031-25G TCIAE1031-100G
2-Ethyl-1-hexanethiol ≥98.0%
2-Ethyl-1-hexanethiol
Formel: C₈H₁₈S
Kokpunkt: 67°C
Flampunkt: 60°C
MDL Number: MFCD00039654
CAS nummer: 7341-17-5
EINECS: 230-854-1
UN: 3336
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR