Ethyl 3-Amino-4-methylbenzoate ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 3-Amino-p-toluic Acid Ethyl Ester, Ethyl 3-amino-4-methylbenzoate, Ethyl 3-Amino-p-toluate, 3-Amino-4-methylbenzoic Acid Ethyl Ester

E1025-5G E1025-25G
TCIAE1025-5GEA 670 SEK
TCIAE1025-5G TCIAE1025-25G
Ethyl 3-Amino-4-methylbenzoate ≥98.0% (by GC, titration analysis)
Ethyl-3-Amino-4-methylbenzoate
Formel: C₁₀H₁₃NO₂
Kokpunkt: 105°C
MDL Number: MFCD01926538
CAS nummer: 41191-92-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 49.0 to 53.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR