Ethyl-1,1,2,2-tetrafluoroethyl ether ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
E1019-5G
E1019-25G
TCIAE1019-5GEA
295
SEK
TCIAE1019-5G
TCIAE1019-25G
Ethyl-1,1,2,2-tetrafluoroethyl ether ≥98.0%
Ethyl-1,1,2,2-tetrafluoroethyl ether
Formel:
C₄H₆F₄O MW: 146,08 g/mol Kokpunkt: 58°C Flampunkt: –15°C |
MDL Number:
MFCD00069162 CAS nummer: 512-51-6 UN: 3271 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...