Ethyl-3-acetoxyhexanoate ≥98.0%

Leverantör: TCI

Synonyms: 3-Acetoxyhexanoic Acid Ethyl Ester

E1014-5G E1014-25G
TCIAE1014-5GEA 575 SEK
TCIAE1014-5G TCIAE1014-25G
Ethyl-3-acetoxyhexanoate ≥98.0%
Ethyl-3-acetoxyhexanoate
Formel: C₁₀H₁₈O₄
MW: 202.25 g/mol
Kokpunkt: 116°C
MDL Number: MFCD00792508
CAS nummer: 21188-61-4
EINECS: 244-263-1

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR