Emtricitabine ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 4-Amino-5-fluoro-1-((2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one

E1007-25MG E1007-250MG
TCIAE1007-25MGEA 215 SEK
TCIAE1007-25MG TCIAE1007-250MG
Emtricitabine ≥98.0% (by HPLC, titration analysis)
Emtricitabine
Formel: C₈H₁₀FN₃O₃S
MW: 247,24 g/mol
MDL Number: MFCD00870151
CAS nummer: 143491-57-0
Merck Index: 14,03565

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 153.0 to 157.0 deg-C
Specific rotation [a]20/D -137.0 to -143.0 deg (C=0.25, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR