Ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: Ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, Ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate

E1005-5G E1005-25G
TCIAE1005-5GEA 2250 SEK
TCIAE1005-5G TCIAE1005-25G
Ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate
Formel: C₉H₁₃NO₂S
MDL Number: MFCD00085051
CAS nummer: 4815-24-1
EINECS: 225-387-5

Order Now

Specifikation testresultat

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 90.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR