4-Ethoxybenzonitrile ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: p-Ethoxybenzonitrile
E0981-5G E0981-25G
TCIAE0981-5GEA 230 SEK
TCIAE0981-5G TCIAE0981-25G
4-Ethoxybenzonitrile ≥98.0%
4-Ethoxybenzonitrile
Formel: C₉H₉NO
MW: 147,18 g/mol
Kokpunkt: 145°C
MDL Number: MFCD00001819
CAS nummer: 25117-74-2
UN: 3439
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 64.0 deg-C