3-(2-Ethylhexyl)thiophene ≥97.0%

Leverantör: TCI EUROPE

E0973-200MG E0973-1G
TCIAE0973-200MGEA 1050 SEK
TCIAE0973-200MG TCIAE0973-1G
3-(2-Ethylhexyl)thiophene ≥97.0%
3-(2-Ethylhexyl)thiophene
Formel: C₁₂H₂₀S
MW: 196,35 g/mol
Kokpunkt: 30°C
Storage Temperature: Kylskåp
MDL Number: MFCD02260336
CAS nummer: 121134-38-1

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR