S-Ethyl thiopropionate ≥98.0%

Leverantör: TCI
Varning

Synonyms: Thiopropionic Acid S-Ethyl Ester

E0971-25ML E0971-5ML
TCIAE0971-25MLEA 1350 SEK
TCIAE0971-25ML TCIAE0971-5ML
S-Ethyl thiopropionate ≥98.0%
S-Ethyl thiopropionate
Formel: C₅H₁₀OS
Kokpunkt: 126°C
Smältpunkt: -95°C
Flampunkt: 36°C
MDL Number: MFCD00027016
CAS nummer: 2432-42-0
EINECS: 219-405-0
UN: 1993
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR