5-Ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine ≥98.0%
Leverantör: TCI EUROPE
E0951-5G
E0951-25G
TCIAE0951-5GEA
765
SEK
TCIAE0951-5G
TCIAE0951-25G
5-Ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine ≥98.0%
5-Ethyl-2-[2-(4-nitrophenoxy)ethyl]pyridine
Formel:
C₁₅H₁₆N₂O₃ MW: 272,30 g/mol |
MDL Number:
MFCD09032965 CAS nummer: 85583-54-6 |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 57.0 to 61.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...