4-Ethoxy-3-nitropyridine ≥98.0%

Leverantör: TCI
Varning

E0942-1G E0942-5G
TCIAE0942-1GEA 550 SEK
TCIAE0942-1G TCIAE0942-5G
4-Ethoxy-3-nitropyridine ≥98.0%
4-Ethoxy-3-nitropyridine
Formel: C₇H₈N₂O₃
MDL Number: MFCD00234959
CAS nummer: 1796-84-5

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR