Ethyl cyclobutanecarboxylate ≥98.0%

Leverantör: TCI EUROPE
Varning

E0931-5G E0931-25G
TCIAE0931-5GEA 550 SEK
TCIAE0931-5G TCIAE0931-25G
Ethyl cyclobutanecarboxylate ≥98.0%
Ethyl cyclobutanecarboxylate
Formel: C₇H₁₂O₂
MW: 128,1690 g/mol
Kokpunkt: 159°C
Flampunkt: 41°C
MDL Number: MFCD00001321
CAS nummer: 14924-53-9
EINECS: 238-995-0
UN: 3272
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR