Ethyl-2-(2-methyl-1,3-dioxolan-2-yl)acetate ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
Ethyl 2-(2-methyl-1,3-dioxolan-2-yl)acetate, 2-(2-Methyl-1,3-dioxolan-2-yl)acetic Acid Ethyl Ester
E0929-25G
E0929-100G
E0929-500G
TCIAE0929-25GEA
230
SEK
TCIAE0929-25G
TCIAE0929-100G
TCIAE0929-500G
Ethyl-2-(2-methyl-1,3-dioxolan-2-yl)acetate ≥98.0%
Ethyl-2-(2-methyl-1,3-dioxolan-2-yl)acetate
Formel:
C₈H₁₄O₄ MW: 174,20 g/mol Kokpunkt: 98°C |
MDL Number:
MFCD00152488 CAS nummer: 6413-10-1 EINECS: 229-114-0 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...