Ethyl-3-bromo-1H-indole-2-carboxylate ≥95.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Ethyl 3-bromoindole-2-carboxylate, Ethyl 3-Bromoindole-2-carboxylate, 3-Bromoindole-2-carboxylic acid ethyl ester, Ethyl 3-bromo-1H-indole-2-carboxylate
E0927-5G
E0927-1G
TCIAE0927-5GEA
1725
SEK
TCIAE0927-5G
TCIAE0927-1G
Ethyl-3-bromo-1H-indole-2-carboxylate ≥95.0%
Ethyl-3-bromo-1H-indole-2-carboxylate
Formel:
C₁₁H₁₀BrNO₂ MW: 268,11 g/mol |
MDL Number:
MFCD02071793 CAS nummer: 91348-45-7 |
Specifikation testresultat
Appearance | White to Amber powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 149.0 to 153.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...