2-((Z)-5-((E)-2-Methyl-3-phenylallylidene)-4-oxo-2-thioxothiazolidin-3-yl)acetic acid ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI
|
Fara
|
E0906-1G
E0906-5G
TCIAE0906-1GEA
310
SEK
TCIAE0906-1G
TCIAE0906-5G
2-((Z)-5-((E)-2-Methyl-3-phenylallylidene)-4-oxo-2-thioxothiazolidin-3-yl)acetic acid ≥98.0% (by HPLC, titration analysis)
2-((Z)-5-((E)-2-Methyl-3-phenylallylidene)-4-oxo-2-thioxothiazolidin-3-yl)acetic acid
Formel:
C₁₅H₁₃NO₃S₂ |
MDL Number:
MFCD00865484 CAS nummer: 82159-09-9 UN: 2811 ADR: 6.1,II Merck Index: 14,03605 |
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 222.0 to 226.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...