Ethyl-2-bromothiazole-4-carboxylate ≥95.0%
Leverantör: TCI EUROPE
Synonyms:
Ethyl 2-bromothiazole-4-carboxylate
E0900-5G
E0900-25G
E0900-1G
TCIAE0900-5GEA
590
SEK
TCIAE0900-5G
TCIAE0900-25G
TCIAE0900-1G
Ethyl-2-bromothiazole-4-carboxylate ≥95.0%
Ethyl-2-bromothiazole-4-carboxylate
Formel:
C₆H₆BrNO₂S MW: 236,08 g/mol Kokpunkt: 154°C Storage Temperature: Kylskåp |
MDL Number:
MFCD03788564 CAS nummer: 100367-77-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 68.0 to 72.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...