β-Estradiol-17-valerate ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: beta-Estradiol 17-Valerate

E0876-1G E0876-5G
TCIAE0876-1GEA 1175 SEK
TCIAE0876-1G TCIAE0876-5G
β-Estradiol-17-valerate ≥98.0%
β-Estradiol-17-valerate
Formel: C₂₃H₃₂O₃
MW: 356,506 g/mol
MDL Number: MFCD00056541
CAS nummer: 979-32-8
EINECS: 213-559-2
Merck Index: 14,03703

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 146.0 to 150.0 deg-C
Specific rotation [a]20/D +41.0 to +44.0 deg (C=2, CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR