N-(Ethoxycarbonyl)nortropinone ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate, 8-(Ethoxycarbonyl)-8-azabicyclo[3.2.1]octan-3-one, N-Carbethoxy-4-tropinone

E0834-25G E0834-5G
TCIAE0834-25GEA 7500 SEK
TCIAE0834-25G TCIAE0834-5G
N-(Ethoxycarbonyl)nortropinone ≥98.0%
N-(Ethoxycarbonyl)nortropinone
Formel: C₁₀H₁₅NO₃
MW: 197,23 g/mol
Kokpunkt: 153°C
MDL Number: MFCD00078171
CAS nummer: 32499-64-2

Order Now

Specifikation testresultat

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR