Ethyl-2-methylvalerate ≥98.0%
Leverantör: TCI
Varning
Synonyms: 2-Methylpentanoic Acid Ethyl Ester, Ethyl 2-methylpentanoate, 2-Methylvaleric Acid Ethyl Ester, Ethyl 2-Methylpentanoate
E0801-25ML E0801-500ML
TCIAE0801-25MLEA 215 SEK
TCIAE0801-25ML TCIAE0801-500ML
Ethyl-2-methylvalerate ≥98.0%
Ethyl-2-methylvalerate
Formel: C₈H₁₆O₂
MW: 144.21 g/mol
Kokpunkt: 153°C
Flampunkt: 39°C
MDL Number: MFCD00036564
CAS nummer: 39255-32-8
EINECS: 254-384-1
UN: 3272
ADR: 3 III
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %