Enrofloxacin ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI
Synonyms:
1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid, 1-Cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
E0786-25G
E0786-5G
TCIAE0786-25GEA
1875
SEK
TCIAE0786-25G
TCIAE0786-5G
Enrofloxacin ≥98.0% (by HPLC, titration analysis)
Enrofloxacin
Formel:
C₁₉H₂₂FN₃O₃ MW: 359.40 g/mol |
MDL Number:
MFCD00792463 CAS nummer: 93106-60-6 Merck Index: 14,03592 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 222.0 to 228.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...