1-Ethyl-3-methylimidazoliumthiocyanate ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
1-Ethyl-3-methyl-1H-imidazol-3-ium thiocyanate
E0776-5G
E0776-25G
TCIAE0776-5GEA
605
SEK
TCIAE0776-5G
TCIAE0776-25G
1-Ethyl-3-methylimidazoliumthiocyanate ≥98.0% (by HPLC, titration analysis)
1-Ethyl-3-methylimidazoliumthiocyanate
Formel:
C₇H₁₁N₃S MW: 169.25 g/mol Smältpunkt: –6°C |
MDL Number:
MFCD06798182 CAS nummer: 331717-63-6 |
|
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...