4-Ethylbenzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI
|
Varning
|
E0773-25G
E0773-5G
TCIAE0773-25GEA
1475
SEK
TCIAE0773-25G
TCIAE0773-5G
4-Ethylbenzenesulphonamide ≥98.0% (by HPLC, total nitrogen)
4-Ethylbenzenesulphonamide
Formel:
C₈H₁₁NO₂S |
MDL Number:
MFCD01365828 CAS nummer: 138-38-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 109.0 to 113.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...