trans-4-Ethylcyclohexanol ≥96.0%

Leverantör: TCI

E0768-5G E0768-1G
TCIAE0768-5GEA 3800 SEK
TCIAE0768-5G TCIAE0768-1G
trans-4-Ethylcyclohexanol ≥96.0%
trans-4-Ethylcyclohexanol
Formel: C₈H₁₆O
Kokpunkt: 84°C
MDL Number: MFCD00070694
CAS nummer: 19781-62-5

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR