3,4-Ethylenedioxythiophene ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 2,3-Dihydrothieno[3,4-b][1,4]dioxine, EDOT
E0741-25G E0741-5G
TCIAE0741-25GEA 1175 SEK
TCIAE0741-25G TCIAE0741-5G
3,4-Ethylenedioxythiophene ≥98.0%
3,4-Ethylenedioxythiophene
Formel: C₆H₆O₂S
MW: 142,17 g/mol
Kokpunkt: 112°C
Flampunkt: 104°C
Storage Temperature: Kylskåp
MDL Number: MFCD02093622
CAS nummer: 126213-50-1
UN: 2810
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %