2-(4-Ethoxyphenyl)-2-methylpropanol ≥98.0%

Leverantör: TCI EUROPE
Varning

E0730-25G E0730-5G
TCIAE0730-25GEA 1900 SEK
TCIAE0730-25G TCIAE0730-5G
2-(4-Ethoxyphenyl)-2-methylpropanol ≥98.0%
2-(4-Ethoxyphenyl)-2-methylpropanol
Formel: C₁₂H₁₈O₂
MW: 194,27 g/mol
MDL Number: MFCD07787279
CAS nummer: 83493-63-4

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 50.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR