2-Ethylpyrrole ≥96.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 2-Ethyl-1H-pyrrole
E0713-1G E0713-5G
TCIAE0713-1GEA 1250 SEK
TCIAE0713-1G TCIAE0713-5G
2-Ethylpyrrole ≥96.0%
2-Ethylpyrrole
Formel: C₆H₉N
MW: 95,15 g/mol
Kokpunkt: 165°C
Flampunkt: 62°C
Storage Temperature: Kylskåp
MDL Number: MFCD00060477
CAS nummer: 1551-06-0
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %