all cis-8,11,14-Eicosatrienoic acid ≥98.0%
Leverantör: TCI EUROPE
Synonyms: Dihomo-gamma-linolenic Acid, Dihomo-γ-linolenic acid, (8Z,11Z,14Z)-Icosa-8,11,14-trienoic acid, all cis-8,11,14-Icosatrienoic Acid
E0640-10MG E0640-50MG
TCIAE0640-10MGEA 550 SEK
TCIAE0640-10MG TCIAE0640-50MG
all cis-8,11,14-Eicosatrienoic acid ≥98.0%
all cis-8,11,14-Eicosatrienoic acid
Formel: C₂₀H₃₄O₂
MW: 306,49 g/mol
Kokpunkt: 200°C
Flampunkt: 62°C
Storage Temperature: Frys
MDL Number: MFCD00065721
CAS nummer: 1783-84-2
EINECS: 217-233-0
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %