3-Ethynylanisole ≥97.0%
Leverantör: TCI EUROPE
Synonyms: 1-Ethynyl-3-methoxybenzene, (3-Methoxyphenyl)ethyne, (3-Methoxyphenyl)acetylene, 1-Methoxy-3-ethynylbenzene
E0627-1G E0627-5G
TCIAE0627-1GEA 615 SEK
TCIAE0627-1G TCIAE0627-5G
3-Ethynylanisole ≥97.0%
3-Ethynylanisole
Formel: C₉H₈O
MW: 132,16 g/mol
Kokpunkt: 90°C
Flampunkt: 77°C
Storage Temperature: Kylskåp
MDL Number: MFCD00160810
CAS nummer: 768-70-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4771a638e1bf4a9cb7346f7d80470bfcf9308896&vid=4771a638e1bf4a9cb7346f7d80470bfcf9308896&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %