N-Ethyl-N-(2-hydroxyethyl)nitrosamine ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: N-Ethyl-N-nitroso-2-aminoethanol

E0580-1G E0580-5G
TCIAE0580-1GEA 710 SEK
TCIAE0580-1G TCIAE0580-5G
N-Ethyl-N-(2-hydroxyethyl)nitrosamine ≥97.0%
N-Ethyl-N-(2-hydroxyethyl)nitrosamine
Formel: C₄H₁₀N₂O₂
MW: 118,14 g/mol
Kokpunkt: 103°C
Storage Temperature: Kylskåp
MDL Number: MFCD01727871
CAS nummer: 13147-25-6

Order Now

Specifikation testresultat

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR