1,4-Epoxy-1,4-dihydronaphthalene ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
1,4-Dihydro-1,4-epoxynaphthalene
E0567-1G
E0567-5G
TCIAE0567-1GEA
1050
SEK
TCIAE0567-1G
TCIAE0567-5G
1,4-Epoxy-1,4-dihydronaphthalene ≥98.0%
1,4-Epoxy-1,4-dihydronaphthalene
Formel:
C₁₀H₈O MW: 144,17 g/mol |
MDL Number:
MFCD00167584 CAS nummer: 573-57-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 55.0 to 59.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...