2-(Ethylamino)pyridine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

E0558-25G E0558-5G
TCIAE0558-25GEA 3875 SEK
TCIAE0558-25G TCIAE0558-5G
2-(Ethylamino)pyridine ≥98.0% (by GC, titration analysis)
2-(Ethylamino)pyridine
Formel: C₇H₁₀N₂
MW: 122,17 g/mol
Kokpunkt: 82°C
MDL Number: MFCD04038418
CAS nummer: 37059-57-7

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR