2-Ethoxy-5-nitropyridine ≥98.0%

Leverantör: TCI
Varning

E0536-25G E0536-5G
TCIAE0536-25GEA 2075 SEK
TCIAE0536-25G TCIAE0536-5G
2-Ethoxy-5-nitropyridine ≥98.0%
2-Ethoxy-5-nitropyridine
Formel: C₇H₈N₂O₃
MDL Number: MFCD01646181
CAS nummer: 31594-45-3

Order Now

Specifikation testresultat

Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR