4-Ethylamino-1-butanol ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-(Ethylamino)butan-1-ol, N-Ethylbutanolamine
E0473-25G
E0473-5G
TCIAE0473-25GEA
2475
SEK
TCIAE0473-25G
TCIAE0473-5G
4-Ethylamino-1-butanol ≥98.0%
4-Ethylamino-1-butanol
Formel:
C₆H₁₅NO MW: 117,19 g/mol Kokpunkt: 86°C |
MDL Number:
MFCD00671534 CAS nummer: 39216-86-9 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...